Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scn5lab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045123.1 | NP_001038588.1 | 7 - 8 | 1013 | 81 | |
GenBank mRNA | DQ149508.1 | - | 7 - 8 | 1013 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |