Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scn5lab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045123.1 | NP_001038588.1 | 9 - 10 | 1354 | 62 | |
GenBank mRNA | DQ149508.1 | - | 9 - 10 | 1354 | 62 | |
DQ837300.1 | - | 10 - 11 | 1707 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Danio rerio Probe spans exons |