Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scn5lab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045123.1 | NP_001038588.1 | 10 - 11 | 1513 | 66 | |
GenBank mRNA | DQ149508.1 | - | 10 - 11 | 1513 | 66 | |
DQ837300.1 | - | 11 - 12 | 1866 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Danio rerio Probe spans exons |