Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mrps18c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045190.2 | NP_001038655.2 | 3 - 4 | 257 | 137 | |
GenBank mRNA | BC134080.1 | - | 3 - 4 | 257 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |