Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mrps18c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045190.2 | NP_001038655.2 | 5 - 6 | 378 | 72 | |
GenBank mRNA | BC134080.1 | - | 5 - 6 | 378 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |