Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nsrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077430.2 | NP_001070898.1 | 2 - 3 | 129 | 65 | |
GenBank mRNA | BC076130.1 | - | 2 - 3 | 149 | 65 | |
BC124124.1 | - | 2 - 3 | 134 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |