Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ctdspl2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077544.1 | NP_001071012.1 | 10 - 11 | 1286 | 62 | |
GenBank mRNA | BC124794.1 | - | 10 - 11 | 1286 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |