Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: il10ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077625.1 | NP_001071093.1 | 3 - 4 | 478 | 118 | |
GenBank mRNA | BC142763.1 | - | 3 - 4 | 585 | 118 | |
EF014957.1 | - | 3 - 4 | 478 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |