Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tyrobp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100103.2 | NP_001093573.2 | 4 - 5 | 354 | 90 | |
XM_009292219.1 | XP_009290494.1 | 4 - 5 | 1327 | 90 | ||
GenBank mRNA | EF158446.1 | - | 4 - 5 | 354 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Danio rerio Probe spans exons |