Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ccdc83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102572.2 | NP_001096042.2 | 1 - 2 | 278 | 130 | |
XM_009305348.3 | XP_009303623.1 | 1 - 2 | 301 | 130 | ||
XM_009305349.3 | XP_009303624.1 | 1 - 2 | 299 | 130 | ||
XM_009305350.3 | XP_009303625.1 | 1 - 2 | 299 | 130 | ||
XM_009305351.2 | XP_009303626.1 | 1 - 2 | 297 | 130 | ||
GenBank mRNA | EF554575.2 | - | 1 - 2 | 278 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons |