Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lmnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131002.2 | NP_571077.2 | 8 - 9 | 1593 | 79 | |
GenBank mRNA | AJ005936.1 | - | 8 - 9 | 1587 | 79 | |
BC162105.1 | - | 8 - 9 | 1447 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |