Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ntn1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131029.2 | NP_571104.1 | 5 - 6 | 1989 | 88 | |
XM_021476776.1 | XP_021332451.1 | 4 - 5 | 1833 | 88 | ||
GenBank mRNA | AF002717.1 | - | 5 - 6 | 1861 | 88 | |
CU638812.1 | - | 5 - 6 | 1861 | 88 | ||
KM655584.1 | - | 5 - 6 | 1482 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Danio rerio Probe spans exons |