Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbx2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131051.1 | NP_571126.1 | 4 - 5 | 1270 | 103 | |
GenBank mRNA | AF136946.1 | - | 4 - 5 | 823 | 103 | |
AF179405.1 | - | 4 - 5 | 1270 | 103 | ||
BC051603.1 | - | 4 - 5 | 1266 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |