Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131294.1 | NP_571369.1 | 4 - 5 | 432 | 122 | |
GenBank mRNA | BC055516.1 | - | 4 - 5 | 432 | 122 | |
BC165134.1 | - | 4 - 5 | 398 | 122 | ||
Y12236.1 | - | 392 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |