Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: anos1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131358.1 | NP_571433.1 | 7 - 8 | 1072 | 95 | |
GenBank mRNA | AF163310.1 | - | 7 - 8 | 1072 | 95 | |
BC060938.1 | - | 7 - 8 | 1076 | 95 | ||
KM655630.1 | - | 7 - 8 | 1025 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |