Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spon1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131515.1 | NP_571590.1 | 11 - 12 | 1640 | 83 | |
GenBank mRNA | AB006086.1 | - | 11 - 12 | 1640 | 83 | |
BC162586.1 | - | 11 - 12 | 1623 | 83 | ||
KM655720.1 | - | 11 - 12 | 1488 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |