Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: st8sia2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153662.1 | NP_705948.1 | 4 - 5 | 1183 | 70 | |
GenBank mRNA | AJ705101.1 | - | 4 - 5 | 573 | 70 | |
AY055462.1 | - | 4 - 5 | 1183 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Danio rerio Probe spans exons |