Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snrpc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173251.2 | NP_775358.1 | 2 - 3 | 95 | 122 | |
GenBank mRNA | AY099526.1 | - | 2 - 3 | 97 | 122 | |
BC091442.1 | - | 2 - 3 | 95 | 122 | ||
BC165527.1 | - | 1 - 2 | 87 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |