Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc34a2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182877.2 | NP_878297.1 | 3 - 4 | 325 | 77 | |
XM_021469757.1 | XP_021325432.1 | 4 - 5 | 775 | 77 | ||
GenBank mRNA | AF297180.2 | - | 3 - 4 | 326 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |