Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vax2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194409.1 | NP_919390.1 | 1 - 2 | 253 | 98 | |
GenBank mRNA | AY183363.1 | - | 1 - 2 | 253 | 98 | |
BC163223.1 | - | 1 - 2 | 345 | 98 | ||
BC163225.1 | - | 1 - 2 | 345 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |