Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: umps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200174.2 | NP_956468.2 | 4 - 5 | 1238 | 106 | |
GenBank mRNA | BC045440.1 | - | 4 - 5 | 1184 | 106 | |
BC055000.1 | - | 4 - 5 | 1184 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |