Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snrnp40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200322.1 | NP_956616.1 | 7 - 8 | 930 | 99 | |
GenBank mRNA | BC051783.1 | - | 7 - 8 | 930 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |