Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: jkamp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200482.2 | NP_956776.2 | 2 - 3 | 174 | 120 | |
GenBank mRNA | BC075939.1 | - | 2 - 3 | 174 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Danio rerio Probe spans exons |