Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gadd45aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200576.2 | NP_956870.2 | 3 - 4 | 464 | 68 | |
GenBank mRNA | AY714218.1 | - | 3 - 4 | 380 | 68 | |
BC056690.1 | - | 3 - 4 | 448 | 68 | ||
BC066476.1 | - | 3 - 4 | 436 | 68 | ||
EH446990.1 | - | 3 - 4 | 473 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Endogenous Controls Recommended Assay Danio rerio Probe spans exons |