Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wdr21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200701.1 | NP_956995.1 | 3 - 4 | 236 | 98 | |
XM_021468379.1 | XP_021324054.1 | 3 - 4 | 273 | 98 | ||
GenBank mRNA | AY423032.1 | - | 3 - 4 | 236 | 98 | |
BC059444.1 | - | 2 - 3 | 189 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |