Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rgs14a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201304.1 | NP_958461.1 | 3 - 4 | 417 | 116 | |
XM_009307644.3 | XP_009305919.2 | 3 - 4 | 383 | 116 | ||
GenBank mRNA | BC055659.1 | - | 3 - 4 | 417 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |