Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hspa9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201326.2 | NP_958483.2 | 14 - 15 | 1761 | 76 | |
GenBank mRNA | BC044175.1 | - | 14 - 15 | 1863 | 76 | |
BC083504.1 | - | 14 - 15 | 1761 | 76 | ||
BC164566.1 | - | 14 - 15 | 1771 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |