Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hspa9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201326.2 | NP_958483.2 | 4 - 5 | 447 | 71 | |
GenBank mRNA | BC044175.1 | - | 4 - 5 | 549 | 71 | |
BC083504.1 | - | 4 - 5 | 447 | 71 | ||
BC164566.1 | - | 4 - 5 | 457 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |