Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nono | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201579.1 | NP_963873.1 | 2 - 3 | 236 | 78 | |
XM_005165105.4 | XP_005165162.1 | 2 - 3 | 618 | 78 | ||
XM_021476008.1 | XP_021331683.1 | 1 - 2 | 149 | 78 | ||
GenBank mRNA | BC046880.1 | - | 2 - 3 | 236 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |