Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mtx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205621.1 | NP_991184.1 | 5 - 6 | 522 | 77 | |
XM_005165510.4 | XP_005165567.3 | 5 - 6 | 585 | 77 | ||
GenBank mRNA | AY194226.1 | - | 5 - 6 | 522 | 77 | |
BC094962.1 | - | 5 - 6 | 522 | 77 | ||
BC164101.1 | - | 5 - 6 | 544 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |