Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ubr7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212629.1 | NP_997794.1 | 6 - 7 | 786 | 113 | |
XM_021468365.1 | XP_021324040.1 | 6 - 7 | 1198 | 113 | ||
GenBank mRNA | BC044390.1 | - | 6 - 7 | 786 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |