Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ethe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212929.1 | NP_998094.1 | 4 - 5 | 763 | 87 | |
GenBank mRNA | BC067574.1 | - | 4 - 5 | 763 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |