Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prdm8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213410.1 | NP_998575.1 | 2 - 3 | 377 | 112 | |
XM_005156810.2 | XP_005156867.1 | 2 - 3 | 517 | 112 | ||
GenBank mRNA | BC055201.1 | - | 2 - 3 | 377 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |