Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rcn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002158.1 | NP_001002158.1 | 3 - 4 | 462 | 93 | |
XM_005164012.4 | XP_005164069.1 | 3 - 4 | 530 | 93 | ||
GenBank mRNA | BC071338.1 | - | 3 - 4 | 462 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |