Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eef1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002371.1 | NP_001002371.1 | 6 - 7 | 1184 | 94 | |
GenBank mRNA | BC075885.1 | - | 6 - 7 | 1184 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |