Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: polr3h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002476.1 | NP_001002476.1 | 2 - 3 | 424 | 101 | |
GenBank mRNA | BC076336.1 | - | 2 - 3 | 424 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |