Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: polr3h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002476.1 | NP_001002476.1 | 5 - 6 | 806 | 128 | |
GenBank mRNA | BC076336.1 | - | 5 - 6 | 806 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |