Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mterf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002615.1 | NP_001002615.1 | 5 - 6 | 850 | 83 | |
GenBank mRNA | BC075975.1 | - | 5 - 6 | 850 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |