Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: adprh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003453.2 | NP_001003453.1 | 3 - 4 | 521 | 92 | |
GenBank mRNA | BC078399.1 | - | 3 - 4 | 367 | 92 | |
BC164170.1 | - | 2 - 3 | 259 | 92 | ||
EH441896.1 | - | 3 - 4 | 519 | 92 | ||
EH557169.1 | - | 3 - 4 | 562 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |