Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: quo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004109.2 | NP_001004109.2 | 21 - 22 | 5968 | 88 | |
GenBank mRNA | AY654285.2 | - | 21 - 22 | 5973 | 88 | |
BC162633.1 | - | 21 - 22 | 5894 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |