Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006059.2 | NP_001006059.2 | 2 - 3 | 295 | 75 | |
XM_005160561.4 | XP_005160618.1 | 2 - 3 | 316 | 75 | ||
GenBank mRNA | BC083265.1 | - | 2 - 3 | 278 | 75 | |
BC164760.1 | - | 2 - 3 | 126 | 75 | ||
DT865424.1 | - | 2 - 3 | 293 | 75 | ||
KM655711.1 | - | 2 - 3 | 116 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |