Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ech1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006068.1 | NP_001006068.1 | 5 - 6 | 684 | 142 | |
GenBank mRNA | BC083254.1 | - | 5 - 6 | 684 | 142 | |
BC152159.1 | - | 5 - 6 | 484 | 142 | ||
BC154279.1 | - | 5 - 6 | 686 | 142 | ||
BC164085.1 | - | 5 - 6 | 502 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |