Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp2n13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007356.1 | NP_001007357.1 | 7 - 8 | 1198 | 91 | |
GenBank mRNA | BC085592.1 | - | 7 - 8 | 1198 | 91 | |
BC164549.1 | - | 7 - 8 | 1214 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |