Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lxn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008596.1 | NP_001008596.1 | 2 - 3 | 279 | 52 | |
XM_005169637.3 | XP_005169694.1 | 2 - 3 | 245 | 52 | ||
XM_021481397.1 | XP_021337072.1 | 2 - 3 | 245 | 52 | ||
GenBank mRNA | BC086837.1 | - | 2 - 3 | 279 | 52 | |
BC164620.1 | - | 1 - 2 | 137 | 52 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |