Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arfrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013289.2 | NP_001013307.1 | 4 - 5 | 448 | 56 | |
XM_005166170.3 | XP_005166227.1 | 4 - 5 | 594 | 56 | ||
GenBank mRNA | BC090265.1 | - | 4 - 5 | 452 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |