Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nadka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017580.1 | NP_001017580.1 | 878 | 104 | ||
GenBank mRNA | BC092723.1 | - | 878 | 104 | ||
BC165884.1 | - | 728 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio |