Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: efcab11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017812.1 | NP_001017812.1 | 1 - 2 | 168 | 124 | |
GenBank mRNA | BC092868.1 | - | 1 - 2 | 168 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |