Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nphs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001018145.2 | NP_001018155.2 | 3 - 4 | 529 | 107 | |
XM_009296250.3 | XP_009294525.1 | 4 - 5 | 806 | 107 | ||
GenBank mRNA | AY956356.1 | - | 3 - 4 | 524 | 107 | |
BC133976.1 | - | 3 - 4 | 529 | 107 | ||
DQ466905.1 | - | 3 - 4 | 504 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |