Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rhof | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020642.1 | NP_001018478.1 | 1 - 2 | 313 | 82 | |
GenBank mRNA | BC095646.1 | - | 1 - 2 | 313 | 82 | |
BC165017.1 | - | 1 - 2 | 165 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |