Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gnpnat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024374.1 | NP_001019545.1 | 3 - 4 | 376 | 114 | |
XM_005169825.4 | XP_005169882.1 | 4 - 5 | 529 | 114 | ||
GenBank mRNA | BC095715.1 | - | 3 - 4 | 376 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |